VWR® MultiTray®/MultiBox® plus, Pipettespisser
Leverandør: VWR Collection
612-2914EA 665 NOK
612-2914 612-2915 612-2599 612-2913 612-2999
VWR® MultiTray®/MultiBox® plus, Pipettespisser
Pipettespisser
Tips racked in MultiTrays®. Compatible with most micropipettes, except 612-2913 which is compatible with Gilson® and Labmate® micropipettes. MultiBox® plus is an autoclavable PP box with dustproof, hinged and sliding lid, which may be opened completely or partially. MultiBox® plus is suitable for use with all MultiTrays®.
  • Tips fit firmly on micropipettor cones, have straight shape for optimum handling, and offer minimum fluid retention
  • MultiBox® plus can be repeatedly autoclaved, but should be placed in an adapted container to avoid contact with the