VWR® Pipette Rack
Leverandør: VWR Collection
612-3848EA 1675 NOK
612-3848
VWR® Pipette Rack
Pipettestativ
Ideal for serological pipettes.
  • Click together horizontally with side connections
  • Securely lock in place with top connections
  • Magnets on both sides of units connect securely and safely to metal surface, such as filling cabinets, fume hoods, refrigerators and freezers
  • Angled shelves store 0,1 to 50 ml pipettes within arm's reach
  • Protective lid encloses the unit to prevent dust build up, and provides an additional shelf for extra space when connected horizontally