Pipettestativ, roterende
Leverandør: KARTELL
261
612-4008EA 2150 NOK
612-4008
Pipettestativ, roterende
Pipettestativ
Rotary pipette holder made from PP, for up to 94 pipettes.
  • Rotating base
  • Accommodates up to 94 pipettes with a diameter not exceeding 14 mm
Ø×H: 230×470 mm