Serological pipettes, Nunc™
Leverandør: Thermo Fisher Scientific
170355
612-4222EA 0 NOK
612-4222
Serological pipettes, Nunc™
Pipetter Serologiske pipetter
PS, sterile.
  • Coloured top, plugged, coloured packaging for easy identification
  • Convenient, black, extra graduations to full pipette volume
  • Non pyrogenic
These accurate, disposable, plastic pipettes are supplied plugged and sterilised.