Serological pipettes, Shortie, Nunc™
Leverandør: Thermo Fisher Scientific
170361
612-4741EA 1200 NOK
612-4741
Serological pipettes, Shortie, Nunc™
Pipetter Serologiske pipetter
PS, transparent, graduated and sterile. These sleek pipettes, made of ultra-clear virgin PS, are ideal for easy manoeuvring under the hood.
  • Disposable, plugged
  • Certified non pyrogenic
  • Bold clear graduations with easy to read scale markings
  • Non drip tip for pipette precision
  • Sterility assurance level SAL 10⁻⁶
Forpakning: Individually wrapped in paper/plastic peel packaging.