VWR® Bulb Pipettes, Two Marks, Class AS
Leverandør: VWR Collection
612-4775EA 265 NOK
612-4775 612-4776 612-4777 612-4778 612-4779 612-4780 612-4781 612-4782 612-4783 612-4784
VWR® Bulb Pipettes, Two Marks, Class AS
Pipetter Fullpipetter
Borosilicate 9.1 glass.
  • Calibrated to deliver (TD, Ex)
  • Ring mark and inscriptions in high contrast amber stain
Sertifiseringer: DIN EN ISO 648
Bestillingsinformasjon: Dated batch identification and batch certificate.