Målekolber, USP, klasse A, blå gradering, BLAUBRAND®
Leverandør: Brand
36953 36954 36938 36947 36948 36945 36943 36949 36951 36952 USP36953 USP36938 USP36943 USP36954 USP36947 USP36949 USP36950 USP36951 USP36952 USP36948 36968 36973 36977 36978 36979 36980 36981 36982 36983 36984 36975 USP36977 USP36975 USP36973 USP36968 USP36945 USP36979 USP36980 USP36978 USP36984 USP36983 USP36982 USP36981 36950
612-4822EA 1300 NOK
612-4822 612-4823 612-4813 612-4816 612-4817 612-4815 612-4814 612-4818 612-4820 612-4821 612-5439 612-5429 612-5430 612-5440 612-5432 612-5434 612-5435 612-5437 612-5438 612-5433 612-5408 612-5409 612-5411 612-5412 612-5413 612-5414 612-5415 612-5416 612-5417 612-5418 612-5410 612-5444 612-5443 612-5442 612-5441 612-5431 612-5446 612-5447 612-5445 612-5451 612-5450 612-5449 612-5448 612-4870
Målekolber, USP, klasse A, blå gradering, BLAUBRAND®
Kolber Målekolber
Borosilicate glass 3,3 with clear glass stopper or PP stopper.
  • Calibrated to contain (TC, In)
  • Narrow neck
  • Marks and inscriptions in high contrast blue enamel
  • DE-M marking
  • With USP batch certificate or with USP individual certificate
DIN EN ISO 1042
Sertifiseringer: DIN EN ISO 1042