VWR®, Transfer Pipettes, Graduated
Leverandør: VWR Collection
612-6803EA 335 NOK
612-6803 612-6805 612-6804 612-6806
VWR®, Transfer Pipettes, Graduated
Pipetter Pasteurpipetter og overføringspipetter
These LDPE disposable, unbreakable 'all in one' pipettes eliminate the potential hazard of broken glass and exposure to infectious materials.
  • Will not shatter or break
  • Non toxic and inert to biological fluids and most acids
  • Can be sealed, refrigerated and gas sterilised
  • No bulb to insert or remove
  • Uniform drop size
They work well whenever there is a need for quick, safe transfer of fluids.
Sertifiseringer: Compliance to US FDA 21 CFR.