Volumetric flasks, class A, blue markings, BLAUBRAND®
Leverandør: Brand
937249 937251 937252 937253 937234 937243 937247 937248 937290 937254 937250 937291 937236 937238 937255 937246 937245 937293 937233
612-3624EA 1275 NOK
612-3624 612-7133 612-3625 612-3626 612-7123 612-3620 612-3622 612-3623 612-7136 612-7134 612-7132 612-7137 BRND937236 612-7124 BRND937255 612-7131 612-7130 612-7138 612-7122
Volumetric flasks, class A, blue markings, BLAUBRAND®
Kolber Målekolber
Borosilicate glass 3.3, with PP stopper.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast blue enamel
  • DE-M marking
  • Individual certificate included
* In addition to DIN.
**Wide neck.
Sertifiseringer: DIN EN ISO 1042