Kompakt byrette, med utskiftbar PTFE-stoppekran, Schellbach-stripe, klasse AS, BLAUBRAND®
Leverandør: Brand
13916 13918 13913
612-7160EA 1625 NOK
612-7160 612-7161 612-7159
Kompakt byrette, med utskiftbar PTFE-stoppekran, Schellbach-stripe, klasse AS, BLAUBRAND®
Byretter
Borosilicate glass 3.3, class AS, DE-M marking.
  • Calibrated to deliver 'TD, Ex'
  • Modular concept - all components, including PTFE stopcock, are replaceable
  • Schellbach stripe
Sertifiseringer: DIN EN ISO 385
Bestillingsinformasjon: Burettes are also available on request with an individual certificate, a USP individual certificate or a DAkkS calibration certificate.
Leveringsinformasjon: Includes burette tube with Schellbach stripe, stopcock with precision tip and batch certificate.