VWR® Pipette Controller
Leverandør: VWR Collection
612-7179EA 4050 NOK
612-7179
VWR® Pipette Controller
Pipetter Pipettefyllere
The VWR® pipette controller is a compact and portable device that features multiple user-selectable modes and speeds to perform a wide range of liquid handling applications. Each device is supplied with a 3-year warranty.
  • Accommodates serological pipettes ranging from 1 to 100 ml
  • 0,45 μm and 0,22 μm replaceable hydrophobic filters available
  • Large LCD display shows speed settings and low battery warning
  • High-capacity Li-ion battery offers over 20 hours of intermittent use (3 hour charge time)
This lightweight, rechargeable pipette controller is idealMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Maks. hastighet 25 ml <5 s