Repetitive pipettes, mechanical, DISTRIMAN®
Leverandør: GILSON
F164001
612-7810EA 3875 NOK
612-7810
Repetitive pipettes, mechanical, DISTRIMAN®
Pipetter Pipetter, repeterene stepper
DISTRIMAN® is a positive-displacement, continuously adjustable repeater-pipette to simplify multiple dispensing when used with Gilson DISTRITIP® syringes.
  • Zero risk of accidental conversion errors with the direct read out
  • Time saving: continuous adjustable volume and numerous distribution of aliquots (up to 125)
  • Faster multiple dispensing
  • Control button for volume selection
  • DISTRITIP® syringes are available in standard and sterilised versions
DISTRIMAN® can dispense aliquots ranging in volume from 1 µl to 1,25 ml with the combination of its threeMore Product Information