Byrette, rett ventilhane, Schellbach, BLAUBRAND®
Leverandør: Brand
12484 12487 12488
612-9958EA 2450 NOK
612-9958 612-9959 612-9960
Byrette, rett ventilhane, Schellbach, BLAUBRAND®
Byretter
Borosilicate glass 3.3, class AS, DE-M marking.
  • Calibrated to deliver 'TD, Ex'
  • Schellbach stripe, ring marks at major graduations
  • Marks and inscriptions in high contrast blue enamel represent the optimum combination of resistance and readability
Lengde: 800 mm
DIN EN ISO 385
Bestillingsinformasjon: Supplied with a batch certificate. Burettes are also available on request with an individual certificate, a USP individual certificate or a DAkkS calibration certificate.