N-Succinimidyl 4-(dimethylamino)benzoate ≥98.0% (by HPLC), LiChropur™ derivatisation reagent, for HPLC, Supelco®

Leverandør: Merck

Synonymer: 1-{[4-(Dimethylamino)benzoyl]oxy}pyrrolidine-2,5-dione

61224-500MG
61224-500MGEA 6310 NOK
61224-500MG
N-Succinimidyl 4-(dimethylamino)benzoate ≥98.0% (by HPLC), LiChropur™ derivatisation reagent, for HPLC, Supelco®
N-Succinimidyl 4-(dimethylamino)benzoate
Formel: C₁₃H₁₄N₂O₄
MW: 262.26 g/mol
MDL nummer: MFCD16875522
CAS-nummer: 58068-85-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80f278569cc88f35ebe2d0154fd7bf7b778cf4e7&vid=396566882d4c8c98d9c1f8f0d4b3f2df671b5704&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR