Pipette stand, rotary
Leverandør: Brand
30900
613-0465EA 1500 NOK
613-0465
Pipette stand, rotary
Pipettestativ
Rotary pipette stand made of PP.
  • For up to 94 graduated or calibrated pipettes of different sizes
  • Rotary model with additional lower support plate to protect pipette tips
  • Slightly raised edge of base retains liquid residues