VWR®, Pipette Tips for Rainin® LTS® Pipettes
Leverandør: VWR Collection
613-0582EA 640 NOK
613-0582
VWR®, Pipette Tips for Rainin® LTS® Pipettes
Pipettespisser
These VWR® pipette tips are made of clear, pre-tested PP resin and fit Rainin® LTS® pipettes.
  • Fit and eject with significantly less force than most conventional pipette tips
  • Ultra-fine points and super-clear resin for excellent viewing of sample
  • Free of RNase, DNase, endotoxins and trace metals
  • Tested for ATP
  • Available in different packaging options
  • Autoclavable
Unlike pre-sterilised pipette tips from other brands, these tips use a validated sterile process (ISO) and can be used for medical research and diagnostics.
Packaging options:
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact,Sustainable Packaging