Pipette guide, 'Guide 300'
Leverandør: DIVERSIFIED BIOTECH
GUIDE-300
613-1451EA 7350 NOK
613-1451
Pipette guide, 'Guide 300'
Well Plate Stands and Well Orienters
This plastic guide enables users to easily target microplate wells using the plastic sliding grid.
  • Reduces eyestrain during tedious pipetting work
  • Suitable for use with multi channel pipettes
  • Adjustable height to accommodate flat or v-bottom micro or PCR plates
The sliding guide is clearly marked with letters for fast and easy orientation. Ideal for long-term pipetting.