VWR® Stepper Pro, Repetitive Pipette
Leverandør: VWR Collection
613-2169EA 3850 NOK
613-2169
VWR® Stepper Pro, Repetitive Pipette
Pipetter Pipetter, repeterene stepper
This pipette is designed for the dispensing of contaminated fluids and volatile liquids. The acceptance of dispenser tips with plunger helps to avoid contamination by ascending aerosols in the tip guaranteeing long-term, secure use.
  • Flexible: Pipette allows 12 tip sizes, 10 volume adjustments and 120 program steps
  • Volume adjustable from 1 to 5500 µl
  • Ideal for long dispensing series due to its low weight (85 g)
  • Ergonomic design allows one handed volume selection, loading and dispensing of liquids
  • Volume adjustments and dosing volumes for different tips are