Syringes, 3-piece, HENKE-JECT®
Leverandør: HENKE SASS WOLF
8300018745
613-2793EA 530 NOK
613-2793
Syringes, 3-piece, HENKE-JECT®
Sprøyter Sprøyter, medisinsk
These syringes are commonly used for embryo transfer and oocyte handling.
  • Rubber piston syringes
  • Double seal for increased safety
  • Pyrogen-free, PVC-free, non-toxic
Syringes have smooth flow, tight and high transparency barrel and the lubricated synthetic rubber piston allows fine dosing.