Automated pipetting assistant, VIAFLO ASSIST
Leverandør: BDP International Inc.
4500
613-3481EA 0 NOK
613-3481
Automated pipetting assistant, VIAFLO ASSIST
Pipetter Pipetter, repeterene stepper
VIAFLO ASSIST together with VIAFLO II electronic pipettes form the perfect symbiosis to prevent users from RSI and to significantly increase the reproducibility of prolonged pipetting protocols such as serial dilutions, plate fillings and reagent additions. Simply mount any VIAFLO II electronic pipette onto the VIAFLO ASSIST pipette adaptor, choose a pipetting protocol and press the RUN button. VIAFLO ASSIST will then obtain the protocol via Bluetooth from the electronic pipette and automatically execute the desired application.
  • Prevents fatigue and repetitive strain injuries (RSI)