VWR® Powerpette Plus, Pipette Controller
Leverandør: VWR Collection
612-3686EA 4175 NOK
612-3686 613-4445
VWR® Powerpette Plus, Pipette Controller
Pipetter Pipettefyllere
Pipette controller for glass and plastic pipettes from 1 to 100 ml.
  • Speed of suction and dispensing controlled through the finger triggers
  • Three aspirate and dispense modes
  • High and low settings both have variable speed
  • Fully charged has approximately 8 hours of continuous use available
  • Low battery light illuminates when 1 hour's power remains
Offers a unique combination of features to give precision fingertip control in a classic streamlined design, suitable for users with large or small hands. Gravity dispense mode is designed for use with 'ToMore Product Information