VWR® Zippette Pro™, Bottle-Top Dispenser
Leverandør: VWR Collection
613-5281EA 3900 NOK
613-5281
VWR® Zippette Pro™, Bottle-Top Dispenser
Flasketoppdispensere
Variable volume, autoclavable.
  • All wetted parts chemically resistant and autoclavable at 121 °C
  • Spring-loaded Zippette Pro requires no priming
  • Easily removable PTFE piston for smooth action and easy cleaning
  • Safety pedestal valve allows the Zippette Pro to be turned off to prevent accidental delivery, whilst ensuring it is drip-free
  • Borosilicate glass barrel, protected with a transparent PP sleeve, can be easily removed and cleaned
Accurate dispensing from an easy to use variable volume bottle-top dispenser. The Zippette Pro has a capacity of 0 to 30More Product Information