Multipacks, Tacta®
Leverandør: Sartorius
LH-729670 LH-729671 LH-729672 LH-729673 LH-729674 LH-729675 LH-729676 LH-729677
613-5626EA 7920 NOK
613-5626 613-5627 613-5628 613-5629 613-5630 613-5631 613-5632 613-5633
Multipacks, Tacta®
Pipetter Pipetter manuelle og elektroniske
The Tacta® Multipacks offer an opportunity to test and get started with Tacta® mechanical pipettes. Tacta® mechanical pipette offers excellent performance and safety in manual pipetting. It is designed to maintain high accuracy and precision in repetitive and long-lasting pipetting series.
  • Reliable results, even over lengthy pipetting periods
  • Controlled and smooth tip ejection
  • The Optilock® system provides flexibility for volume adjustment and locking
There are different multipack options to choose from ranging from a set of 3 to a set of 5 pipettes.