VWR® Pipette Charging Stand, Rotary
Leverandør: VWR Collection
613-5747EA 6775 NOK
613-5747
VWR® Pipette Charging Stand, Rotary
Pipettestativ
This power carousel is a comfortable charging solution for up to five electronic pipettes.
  • Protects valuable instruments from shocks and contamination
  • Pipettes hang vertically to prevent liquids from running inside the shaft
  • Rotating head gives users easy access to the pipette of their choice
  • Slim design allows users to have a neat, tidy and organised work bench
  • Unrollable cable will avoid any cable spaghetti issue