VWR® Ultra-High Performance, Multi-channel pipettes, mechanical, variable volume
Leverandør: VWR Collection
613-1498EA 12900 NOK
613-1498 613-7070 613-7071 613-7072 613-7073 613-7074 613-7075 613-7076 613-7077
VWR® Ultra-High Performance, Multi-channel pipettes, mechanical, variable volume
Pipetter Pipetter manuelle og elektroniske
VWR® Ultra-High Performance pipettes offer superior accuracy and precision, with proven ergonomic, lightweight and durable design.
  • Low plunger forces
  • Universal tip capability
  • Automatic locking volume adjustment wheel
  • Levered tip ejection and colour-coded for volume identification
  • Easy in-house calibration and maintenance
  • Fully autoclavable