Pipette stand, linear
Leverandør: Sartorius
725620
613-8706EA 1575 NOK
613-8706
Pipette stand, linear
Pipettestativ
This pipette stand holds pipettes in an upright position while not in use to avoid contamination from benches and tables. Designed for all Sartorius mechanical and electronic pipettes, particularly for mLINE®, Proline® Plus and Proline® mechanical devices. Also compatible with other brands.