Salt tester for dairy products
Leverandør: GERBER FUNKE
4760 4770
614-0194EA 2425 NOK
614-0194 614-0195
Salt tester for dairy products
Mat og drikkevare testsystemer
Titration method with Dr Schilling pattern burettes and brown PE bottle.