Reference dryer RD-8
Leverandør: GERBER FUNKE
5700
614-0200EA 0 NOK
614-0200
Reference dryer RD-8
Termostatblokker Digitale termostatblokker
The RD-8 was specifically developed in conjunction with COKZ (the Netherlands Controlling Authority for Milk and Milk products) for determining the moisture content of dried milk in accordance with ISO 5537, IDF 26. Up to eight samples can be tested simultaneously under accurately controlled temperature and pressure.
  • Temperature is regulated by a PID system, and four PT100 sensors determine the temperature
  • Constant pressure regulator maintains the airflow at 33 ml/min within the 8 drying capillaries
NB: *RT = omgivelse