Milk hydrometer with thermometer
Leverandør: Alla France
2200TP018/20QP
614-1501EA 1425 NOK
614-1501
Milk hydrometer with thermometer
Aerometere
This Dornic milk hydrometer with built-in thermometer is graduated from 18 to 38 (1018 to 1038).
  • Internal thermometer and temperature correction indicator
  • Control thermometers are also available
NOTE: Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.