Falling ball viscometer
Leverandør: HAAKE
356-0001
616-3282EA 49975 NOK
616-3282
Falling ball viscometer
Viskosimetere Viskosimeter, fallende ball
This falling ball viscometer provides an accurate way of measuring the viscosity, in transparent Newtonian liquids and gases.
Specially suited for the chemical (polymer solutions, solvents, inks), pharmaceutical (raw materials, glycerine), food (gelatin, sugar solutions) and mineral oil (oils, liquid hydrocarbons) industries.
Sertifiseringer: DIN 53015, ISO 12058
Leveringsinformasjon: Supplied with six balls, instrument case, thermometer, cleaning tools, calibration sheet and instruction manual.