Nedsenkningsstrømkopper
Leverandør: Industrial Physics
VF2071 VF2072 VF2073 VF2074 VF2075 VF2077
616-3412EA 2100 NOK
616-3412 616-3413 769-1144 616-3414 616-3415 616-3417
Nedsenkningsstrømkopper
Viskosimetere Viskosimeter med flytekopp
Dip cups can be used to provide a quick 'break-thread' flow time comparative measurements on site in a manufacturing environment.
  • Titanium anodised aluminium with fixed stainless steel nozzle and handle
  • Engraved with a unique serial number
  • Easy to clean
Sertifiseringer: DIN 53211
Bestillingsinformasjon: The calibration certificate needs to be ordered separately.
Leveringsinformasjon: Stand is not included.