Nα-(p-Toluenesulphonyl)-DL-lysine chloromethyl ketone hydrochloride ≥95% (by elemental analysis), off-white solid, Calbiochem®

Leverandør: Merck Millipore Calbiochem

616382-50
616382-50EA 1325 NOK
616382-50
Nα-(p-Toluenesulphonyl)-DL-lysine chloromethyl ketone hydrochloride ≥95% (by elemental analysis), off-white solid, Calbiochem®
Nalpha-(p-Toluenesulphonyl)-DL-lysine chloromethyl ketone hydrochloride

Inhibits trypsin-like serine proteinases.


Irreversibly inactivates trypsin without affecting chymotrypsin. Prevents nitric oxide production by activated macrophages by interfering with transcription of the iNOS gene. Blocks cell-cell adhesion and binding of HIV-1 virus to the target cells. In macrophages, blocks nitric oxide synthase induced by interferon-γ and lipopolysaccharides (EC₅₀=80 µM). Prevents endonucleolysis accompanying apoptotic death of HL-60 leukemia cells and normal thymocytes.

Formel: C₁₄H₂₂Cl₂N₂O₃S
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00012513
CAS-nummer: 4238-41-9

Bestill nå

Spesifikasjon Testresultater

Purity (Elemental Analysis) ≥95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR