Thermometer rack, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
F189810001
620-0099EA 1725 NOK
620-0099
Thermometer rack, SP Bel-Art
Termometerholdere
White PP. Stores up to 25 thermometers.
  • Supports thermometers vertically
  • Minimises any risk of breakage
  • Clearly organises your storage
  • Rapid access to the thermometers