Temperature chart recorders, disposable
Leverandør: DICKSON
D302
620-0432EA 1775 NOK
620-0432
Temperature chart recorders, disposable
Punktskriver Skrivere
These are single use logger accurately records temperatures from start to finish of transport and documents adherence to critical storage environments.
  • Failsafe start-up and pull out start tab
  • Tamper-proof crimped seal
  • Shock and vibration protection
Leveringsinformasjon: Supplied with a shipment/job sheet and a mounting tape.