VWR® Enviro Safe Dry Block/Incubator Thermometers
Leverandør: VWR Collection
620-0790EA 930 NOK
620-0790 620-0791
VWR® Enviro Safe Dry Block/Incubator Thermometers
Termometere Analogue Thermometers
Designed for use in all standard heating and drying blocks, water baths and incubators.
  • Partial immersion
  • Individual serial numbers
  • PFA coating on 620-0791 helps prevent liquid and glass in the event of breakage
  • Same accuracy as similar mercury thermometers, but without the hazards
Contains enviro safe, non toxic, non hazardous, green liquid against white back glass.
Sertifiseringer: Traceable to NIST; hand inspected in triple-accredited/-registered ISO/IEC 9001:2008, ISO/IEC 14001:2004, ISO/IEC 17025:2005 facility and A2LA Accredited CalibrationMore Product Information