VWR® Enviro-Safe®, Environmentally Friendly Precision Liquid-in-Glass Thermometer
Leverandør: VWR Collection
620-0801EA 3300 NOK
620-0801
VWR® Enviro-Safe®, Environmentally Friendly Precision Liquid-in-Glass Thermometer
Termometere Analogue Thermometers
This high precision, EnviroKleen™ certified thermometer is accurate to ±1,0 °C and features green, non toxic, enviro safe liquid against lead-free, white back glass.
  • Lead-free glass
  • Accuracy equal to similar mercury filled thermometers
  • Individual serial numbers
Suitable for use in the chemical industry, universities, scientific institutions, food and beverage processing, petrochemical, pharmaceutical and medical industries, manufacturers, dairies, breweries and distilleries.
Traceable to NIST/DKD.
Sertifiseringer: Traceable to NIST.