VWR®, Presisjonstermometre, frio temp
Leverandør: VWR Collection
620-0837EA 2175 NOK
620-0837 620-0838 620-0839 620-0840 620-0841
VWR®, Presisjonstermometre, frio temp
Termometere Analogue Thermometers
For kontroll i frysere, kjøleskap, inkubatorer, ovner og varmeskap. Stavtermometer montert i flaske.
  • Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample
  • Insert the thermometer into the bottle and place near the samples for best results
  • Thermometers contain organic liquid; feature permanently fused markings and lead free glass
  • PFA coating safely contains glass and liquid in case of breakage
  • Individuelt serienummer
Insert the thermometer into the bottle and place near theMore Product Information