VWR®, Ultra-low liquid-in-glass thermometers
Leverandør: VWR Collection
620-0857EA 1900 NOK
620-0857 620-0858 620-0860 620-1927 620-1928
VWR®, Ultra-low liquid-in-glass thermometers
Termometere Analogue Thermometers
These thermometers meet all requirements for physical dimensions and accuracy.
  • Meniscus visible in the appropriate low temperature application
  • Possible magnification for more readability
  • White background, fused markings
  • Designed for ultra-low temperature use
  • Feature mercury-free liquid against lead-free, white back glass
Division: 1,0 °C
Accuracy: ±2,0 °C
Traceable to NIST/DKD. Compliant with ANSI/SAMA Z236.1-1983.
Sertifiseringer: Traceable to NIST. Compliant with ANSI/SAMA Z236.1-1983.
Leveringsinformasjon: Supplied with a multi-languageMore Product Information