VWR®, Precision Thermometers, Liquid-in-Glass
Leverandør: VWR Collection
620-0885EA 8875 NOK
620-0885
VWR®, Precision Thermometers, Liquid-in-Glass
Termometere Analogue Thermometers
These liquid-in-glass thermometers feature blue organic liquid fill against lead-free white back glass. They are ideal for use in chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical/medical industry, manufacturing, dairies, breweries and distilleries.
  • Fused markings
  • Individually calibrated and serialised
  • Graduated with fractional degrees
Traceable to NIST/DKD.
Sertifiseringer: Traceable to NIST.
Leveringsinformasjon: Supplied with individual thermometerMore Product Information