Digitaltermometer, TTX 100 Serien
Leverandør: EBRO
1340-5110
620-1384EA 1075 NOK
620-1384
Digitaltermometer, TTX 100 Serien
Termometere Digitaltermometer
Thermocouple thermometer with fixed probe (Type T).
  • Fast measurements, high accuracy
  • HACCP conform
  • Robust and impact resistant
  • Replaceable battery
  • Automatic switch-off
  • HOLD function
Ideal for use in the food industry, transport and storage applications.
EN 13485, conform to HACCP
Sertifiseringer: Conforms to HACCP and EN 13485.
Leveringsinformasjon: Supplied with a replaceable battery and factory calibration certificate.