Infrarøde termometre, TFI 550 og 650
Leverandør: EBRO
1340-1786
620-1553EA 3400 NOK
620-1553
Infrarøde termometre, TFI 550 og 650
Termometere Digitaltermometer
Infrared thermometers, suitable for measurements wherever direct contact is impossible or impractical. With NiCr-Ni connection.
  • Double-laser pointer
  • Alarm when MIN/MAX exceeded
  • Protection class IP 20
  • Factory calibration certificate
  • Battery: 2× AAA (micro)
  • Battery lifetime: typically 140 hours
  • Distance: spot ratio for TFI 550 model =30:1 / spot ratio for TFI 650 model=50:1