Core thermometer, TDC 150
Leverandør: EBRO
1340-1611
620-1568EA 700 NOK
620-1568
Core thermometer, TDC 150
Termometere Digitaltermometer
Core thermometer works with fast response time.
  • Fixed probe, pointed
  • Robust and impact-resistant
  • Replaceable battery
  • Switchable between °C/°F