VWR® Traceable® Alarm Thermometer with Timer
Leverandør: VWR Collection
620-1662EA 805 NOK
620-1662
VWR® Traceable® Alarm Thermometer with Timer
Termometere Digitaltermometer
This thermometer, with timer, is designed to monitor liquids, air, gas or semi-solids. The metal probe may be placed in ovens and incubators making this unit ideal for use in these environments.
  • Display shows time, current temperature and alarm setting
  • Metal probe for applications in ovens and incubators
  • Adjustable display and large digits
Curved stainless steel probe (Ø×L: 4×215 mm). The cable is made of plastic.
Sertifiseringer: Traceable to NIST. Compliant with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.