VWR® Traceable® Digital Thermo-Hygrometer
Leverandør: VWR Collection
620-1673EA 1100 NOK
620-1673
VWR® Traceable® Digital Thermo-Hygrometer
Hygrometere Digitale termohygrometere
This thermometer/clock/humidity monitor shows time, temperature and humidity simultaneously. Internal sensors for use in hoods and storerooms.
  • Memory-clear function
  • Maximum/minimum memory
  • Memory recalls the highest and lowest readings
W×D×H: 59×108×15 mm
Certificate indicates traceability to standards provided by NIST.
Sertifiseringer: Certificate indicates traceability to standards provided by NIST. Calibration compliant with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.
Leveringsinformasjon: Supplied with Traceable® certificate of calibration,More Product Information