VWR® Traceable®, Triple-Display Thermometers
Leverandør: VWR Collection
620-1822EA 2350 NOK
620-1822 620-1823 620-1827
VWR® Traceable®, Triple-Display Thermometers
Termometere Digitaltermometer
These highly accurate thermometers monitor temperatures simultaneously in refrigerators and freezers, two refrigerator locations. Temperature-buffered bottle sensor eliminates transient temperature changes when refrigerator door is opened. Patented unit fulfils all CDC thermometer and vaccine thermometer requirements. They may also be used in water baths, heating blocks and incubators.
  • Displays exact time and date when an alarm is triggered, alarms are programmable in 0,1 °C increments
  • Monitor minimum and maximum readings and displays exact time and date