VWR® Traceable®, Big Digit Alarm Thermometer
Leverandør: VWR Collection
620-1858EA 780 NOK
620-1858
VWR® Traceable®, Big Digit Alarm Thermometer
Termometere Digitaltermometer
This digital thermometer allows the simultaneous reading of ambient and probe temperatures. Audible alarm sounds at high/low temperatures settings, between two settings and outside two settings. Green/red lights and alarm activates when 'out of range' condition is detected.
  • Dual display
  • Jumbo digits
  • Reset at the touch of a button
External probe (L×Ø: 16×5 mm) with 1 metre cable.
Leveringsinformasjon: Supplied with Traceable certificate of calibration, AAA alkaline battery, suction cup for mounting sensor and plastic case.