Infrared printer
Leverandør: Testo
0554.0549
620-1903EA 3675 NOK
620-1903
Infrared printer
Printere Document Printers
The universal IR printer completes data transfers in two seconds. This printer allows users to evaluate restoration measures of air moisture, material equilibrium moisture, and pressure dew point quickly and without error.
  • Hands-free operation
  • Wireless infrared interface for fast data transfer
  • Provides fast, easy to read printouts of measurements
  • Energy saving through auto-off feature
  • Instrument memory for up to 10000 readings
Leveringsinformasjon: Printer supplied wih a wireless infrared interface, one roll of thermal paper, and four AA batteries.