Dual infrared and core thermometer HACCP, testo 104-IR
Leverandør: Testo
0560.1040
620-1904EA 2100 NOK
620-1904
Dual infrared and core thermometer HACCP, testo 104-IR
Termometere Digitaltermometer
This waterproof folding thermometer is a handy aid when carrying out precise food monitoring measurements at any time. It is suitable for applications in production and storage. After use, the probe can be folded away, and the instrument stored safely until the next measurement.
  • Automatic final value recognition thanks to auto hold
  • Storage of minimum/maximum values
  • Rubber coated surface guarantees non-slip handling
* Infrared
** NTC
Weight: 165 g
Compliant with Hazard Analysis and Critical Control Points (HACCP) and EN 13485.
Sertifiseringer: More Product Information