VWR®, Digital Refrigerator and Freezer Thermometer
Leverandør: VWR Collection
620-1913EA 525 NOK
620-1913
VWR®, Digital Refrigerator and Freezer Thermometer
Termometere Digitaltermometer
Digital thermometer for refrigerators and freezers.
  • High and low alarm setting
  • Waterproof fridge sensor
  • °C/°F exchange
Calibration points: 0 and 50 °C
Sertifiseringer: This thermometer comes with a Traceable certificate.
Leveringsinformasjon: Supplied with sensor holder, battery and VELCRO® magic tape.