VWR® Traceable®, Infrared Thermometer
Leverandør: VWR Collection
620-1924EA 3475 NOK
620-1924
VWR® Traceable®, Infrared Thermometer
Termometere Digitaltermometer
This dual-laser (IR) digital thermometer with (K) probe is suitable for non invasive measurements in food, life sciences, pharmaceuticals, petroleum products, cleanrooms, electronics and on-site applications.
  • Field of view of 12:1 (the spot size is 50 mm at 0,60 m)
  • Readings on any type of surface: Solids, semi-solids and liquids
  • Minimum/maximum features for viewing at any time, display of the average of all readings and of the difference between two readings
  • Audible, user-settable, high and low alarms signal out of range conditions
  • User adjustable